missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPNS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113145
This item is not returnable.
View return policy
Description
CAPNS2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The Calpain family of proteins are calcium-regulated thiol proteases which have broad endopeptidase activity throughout the body. Calpain small subunit 2, also known as CSS2 or CAPNS2, is a calcium-dependent protease that is expressed ubiquitously in the cytoplasm. Part of a heterodimer composed of a small subunit and a large subunit, CSS2 catalyzes proteolysis of various proteins involved in cytoskeletal remodeling and signal transduction. CSS2 also acts as a chaperone to the larger subunit, mediating its correct folding and conformation. When bound as a heterodimer, CSS2 is thought to keep the catalytic activity of the large subunit dormant. After binding calcium, CSS2 is released from the complex, thereby activating the large subunit and allowing CSS2 to translocate from the cytoplasm to the cell membrane. Defects in the gene encoding CSS2 result in incorrect calpain activity and retarded fetal development, suggesting that CSS2 expression is essential for proper growth.
Specifications
| CAPNS2 | |
| Polyclonal | |
| Unconjugated | |
| CAPNS2 | |
| 2310005G05Rik; 30K-2; calcium-dependent protease small subunit 2; Calpain small subunit 2; calpain, small subunit 2; CAPNS2; CSS2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 69543, 84290 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96L46, Q9D7J7 | |
| CAPNS2 | |
| Recombinant Human Calpain small subunit 2 protein (1-248AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction