missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPN2 Mouse, anti-Human, CoraLite Plus 594, Clone: 1E1F10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | CAPN2 |
|---|---|
| Clone | 1E1F10 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18089315
|
Proteintech
CL594-66977-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098035
|
Proteintech
CL594-66977-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CAPN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAPN2. This antibody recognizes human, mouse, rat antigen. The CAPN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 594.
Specifications
| CAPN2 | |
| 1000 μg/mL | |
| CoraLite Plus 594 | |
| Mouse | |
| 824 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1E1F10 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| Calpain 2 catalytic subunit, Calpain 2 large subunit, Calpain large polypeptide L2, Calpain M type, CANP 2, CANP2, CANPL2, CANPml, CAPN2, FLJ39928, M calpain, mCANP, Millimolar calpain | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title