missing translation for 'onlineSavingsMsg'
Learn More

Capmatinib, MedChemExpress

Product Code. 16425668
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425668

Brand: Medchem Express HY13404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Capmatinib (INC280; INCB28060) is a potent, orally active, selective, and ATP competitive c-Met kinase inhibitor (IC50=0.13 nM).

Capmatinib can inhibit phosphorylation of c-MET as well as c-MET pathway downstream effectors such as ERK1/2, AKT, FAK, GAB1, and STAT3/5. Capmatinib potently inhibits c-MET-dependent tumor cell proliferation and migration and effectively induces apoptosis. Antitumor activity. Capmatinib is largely metabolized by CYP3A4 and aldehyde oxidase.

Specifications

Chemical Name or Material Capmatinib
CAS 1029712-80-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H17FN6O
Quantity 10 mg
Synonym INC280 INCB28060
Solubility Information DMSO : 25 mg/mL (60.62 mM; Need ultrasonic) ∣H2O : 10 mg/mL (24.25 mM; Need ultrasonic)
SMILES O=C(NC)C1=CC=C(C2=NN3C(N=C2)=NC=C3CC4=CC=C5N=CC=CC5=C4)C=C1F
Molecular Weight (g/mol) 412.42
Formula Weight 412.42
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.