missing translation for 'onlineSavingsMsg'
Learn More

Capivasertib, MedChemExpress

Product Code. p-200096128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16469097

Brand: Medchem Express HY15431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, respectively.

Capivasertib (AZD5363) is an orally active and potent pan-AKT kinase inhibitor with IC50 of 3, 7 and 7 nM for Akt1,Akt2 and Akt3, respectively.

Specifications

Chemical Name or Material Capivasertib
CAS 1143532-39-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25ClN6O2
Quantity 5 mg
Synonym AZD5363
Solubility Information DMSO : 125 mg/mL (291.43 mM; Need ultrasonic)
SMILES O=C(C1(N)CCN(C2=C3C(NC=C3)=NC=N2)CC1)N[C@H](C4=CC=C(Cl)C=C4)CCO
Molecular Weight (g/mol) 428.92
Formula Weight 428.92
Percent Purity 95.09%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.