missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps

Click to view available options
Color:
Blue
Green
Red
White
Yellow
No. per Case:
2000/Cs.
Unit Size:
Pack of 2000
Description
Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.
- For 10 and 12 mL serum transport tubes
Specifications
Specifications
| Diameter (Metric) Outer | 15 mm |
| Product Type | Screw Cap |
| Color | Green |
| No. per Case | 2000/Cs. |
| Quantity | 2000/Cs. |
| For Use With (Equipment) | Capitol Vial 10 and 12 mL Transport Tubes (not included) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction