Learn More
Thermo Scientific™ Thermo Scientific™ Split Specimen Urine Collection Kit
GREENER_CHOICE
Product Code. 15698568
Change view
Click to view available options
No. per Case:
100/Cs.
Unit Size:
Pack of 100
Product Code. No. per Case unitSize
15698568 100/Cs. Pack of 100
1 options
Product Code. 15698568 Supplier Thermo Scientific™ Supplier No. 92023
Perform sample collection and storage with these capitol vial split foil top specimen collection kits.
Perform sample collection and storage with the Thermo Scientific™ Split Foil Top Specimen Collection Kit. It is ideal for SAMHSA workplace testing
Features:
  • Easy-to-use kit—simply peel away the foil top lid, remove the components and provide donor with the collection cup
  • After sample collection, pour 30mL of urine into the first transport vial and 15mL into the second vial, close the hinged lid until it snaps
  • Place vial in the front pocket of specimen bag and completed requisition in the rear pocket
  • Remove the blue release liner on the bag and fold flap over the pockets to seal
Ordering Information:
Custom branding available for foil tops and specimen bags.
Includes:
Beaker (8 oz.) with temperature indicator strip, two (2) 45mL custody vials, generic dual-pocket specimen bag, absorbent foil top.
Note:
For testing according to Substance Abuse and Mental Health Services Administration guidelines, refer to SAMSHA instruction for collection and transport requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection Kit
Material Polypropylene
Sterility Non-sterile
Description Split Foil Top Specimen Collection Kit, 100/Cs
For Use With (Application) U.S. Federal Government Worplace Drug Testing Programs
Includes 8 oz. beaker with temperature indicator strip, 2 x 45 mL Transport Tubes, a generic dual pocket specimen bag with absorbent and a foil top seal.
No. per Case 100/Cs.