missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Capillary Cutter, 310 Genetic Analyzer

Product Code. 15839726 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839726 2 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839726 Supplier Applied Biosystems™ Supplier No. 401958

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a capillary cutter for use with 310 genetic analyzer capillaries. Shortened capillaries can be used for short run modules.

This is a capillary cutter for use with 310 Genetic Analyzer capillaries. Shortened capillaries can be used for short run modules.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Capillary Cutter
Quantity 2 Each
Shipping Condition Room Temperature

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.