Learn More
Macherey Nagel Capillary column OPTIMAWAXplus ID: 0.25 mm, film thickness: 0.50 µm
Brand:  Macherey Nagel 726381.30
Capillary column OPTIMAWAXplus ID: 0.25 mm, film thickness: 0.50 µm
Product Code. 12793371
  • £708.00 / Pack of 1
Only null left
Estimated Shipment: 01-06-2026