missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capicua Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-59906CL1
This item is not returnable.
View return policy
Description
Capicua Polyclonal antibody specifically detects Capicua in Canine,Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Gene Knock-Out,Immunocytochemistry/Immunofluorescence
Specifications
| Capicua | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Purified |
| Immunoprecipitation, Western Blot, Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23152 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction