missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPG Recombinant Rabbit Monoclonal Antibody (5V9G1)

Brand: Thermo Scientific MA538117
This item is not returnable.
View return policy
Description
Specifications
| CAPG | |
| Recombinant Monoclonal | |
| 0.54 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24452, P40121, Q6AYC4 | |
| CAPG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-188 of human CAPG (P40121). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5V9G1 | |
| Unconjugated | |
| CAPG | |
| Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12332, 297339, 822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction