missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CapG Mouse anti-Human, Mouse, Clone: 1D10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59450-100UL
This item is not returnable.
View return policy
Description
CapG Monoclonal specifically detects CapG in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CapG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1054, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1D10 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| CAPG | |
| Recombinant human CAPG (1-348aa) purified from E. coli | |
| 100 μL | |
| Cytoskeleton Markers | |
| 822 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction