missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPG Monoclonal Antibody (K52030_15B12), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | CAPG |
|---|---|
| Clone | K52030_15B12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30201439
|
Invitrogen™
MA555907 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211996
|
Invitrogen™
MA556287 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RDLALAIRDS ERQGKAQVEI VTDGEEPAEM IQVLGPKPAL KEGNPEEDLT ADKANAQAAA LYKVSDATGQ MNLTKVADSS
CAPG Monoclonal antibody specifically detects CAPG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CAPG | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Actin regulatory protein CAP-G; actin-capping protein GCAP39; actin-regulatory protein CAP-G; AFCP; Capg; capping actin protein, gelsolin like; capping protein (actin filament), gelsolin-like; epididymis secretory protein Li 66; gCap39; gelsolin-like capping protein; HEL-S-66; macrophage capping protein; macrophage-capping protein; mbh1; MCP; myc basic motif homolog 1 | |
| Recombinant human CAPG (immunogen range is 191-270). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CAPG |
| K52030_15B12 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P40121 | |
| 822 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title