missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CapG Antibody (1D10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59450CL1
This item is not returnable.
View return policy
Description
CapG Monoclonal antibody specifically detects CapG in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CapG | |
| Monoclonal | |
| CoraFluor 1 | |
| actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
| Recombinant human CAPG (1-348aa) purified from E. coli | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 822 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1D10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction