missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPC/ LRRC26 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81900
This item is not returnable.
View return policy
Description
CAPC / LRRC26 Polyclonal specifically detects CAPC / LRRC26 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CAPC / LRRC26 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| LRRC26 | |
| LRRC26 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human LRRC26. The immunogen is located within the last 50 amino acids of LRRC26. Amino Acid Squence: ALRPPRPPDPNPDPDPHG | |
| Affinity Purified | |
| RUO | |
| 389816 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| CAPC, cytokeratin associated protein, leucine rich repeat containing 26 | |
| Rabbit | |
| 48 kDa | |
| 0.1 mg | |
| Primary | |
| CAPC/LRRC26 antibody is human specific. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction