missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAP1 Recombinant Rabbit Monoclonal Antibody (23GB3020), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£179.00 - £485.00

Specifications

Antigen CAP1
Clone 23GB3020
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089347
GREENER_CHOICE
View Documents
Invitrogen™
MA553318
100 μL
£485.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088547
View Documents
Invitrogen™
MA553319
20 μL
£179.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CAP1 Recombinant Monoclonal antibody specifically detects CAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

CAP1
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q01518
10487
IgG
Affinity chromatography
Antibody
23GB3020
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p
A synthesized peptide derived from CAP1 (12-45AA).
Primary
-20°C
CAP1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.