missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Cap

Product Code. 11259939 Shop All Miele Products
Change view
Click to view available options
:
Size: 4mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11259939 Size: 4mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11259939 Supplier Miele Professional™ Supplier No. 656911

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Spare part for Lab washer G 7835 CD, G 7836 CD, PG 8535, PG 8536

Cap for a nozzle 4mm
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cap
Color Black
Diameter (Metric) 4 mm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.