missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Replacement 2in. Wash Bottle Cap

Product Code. 14885661
Click to view available options
Color:
Red
This item is not returnable. View return policy
This item is not returnable. View return policy

Keep several on hand in case caps are lost or damaged. SP Scienceware™ Replacement 2in. Wash Bottle Cap are color coded caps for wide-mouth wash bottles.

  • Replaceable caps that fits wash bottles with a 53mm (2 in.) mouth
  • A variety of colors for color-coding of wash solutions or applications
  • Durable polypropylene caps with bent washing nozzles

Specifications

Color Red
No. per Case 1 Cs.
Product Type Wash Bottle Closure
For Use With (Equipment) 53mm Mouth wash bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'