missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Canthaxanthin

Product Code. 16408836
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408836 Supplier MedChemExpress Supplier No. HYB1960
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Canthaxanthin is a red-orange carotenoid with various biological activities, such as antioxidant, antitumor properties.

Specifications

Content And Storage -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Molecular Weight (g/mol) 564.84 g/mol
Shipping Condition Shipping with dry ice
For Use With (Application) Cancer-programmed cell death
CAS 514-78-3
Product Type Natural Products
Purity 95.0%
Solubility DMSO : 1 mg/mL (1.77 mM∣ultrasonic and warming and heat to 80°C) ∣H2O : 1 mg/mL (1.77 mM∣ultrasonic and warming and heat to 80°C)
Synonym E 161g∣ all-trans-Canthaxanthin
Target Reactive Oxygen Species∣Endogenous Metabolite
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.