Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids (UKAS ISO 17025 certified)
Product Code. 15969989
Change view
Click to view available options
Viscosity Range:
0.5 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 250 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
15969989 0.5 to 2 cSt Each
15989989 0.8 to 4 cSt Each
15909999 1.6 to 8 cSt Each
15929999 3 to 15 cSt Each
15949999 7 to 35 cSt Each
15969999 20 to 100 cSt Each
15989999 50 to 250 cSt Each
16000000 100 to 500 cSt Each
16020000 240 to 1200 cSt Each
16040000 500 to 2500 cSt Each
16060000 1600 to 8000 cSt Each
16080000 4000 to 20,000 cSt Each
16000010 10,000 to 50,000 cSt Each
16020010 20,000 to 100,000 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15969989 Supplier Paragon Scientific™ Supplier No. CFR01UKC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
  • Supplied with full UKAS ISO 17025 calibration
Specifications
Viscosity Range 0.5 to 2 cSt
Calibrated Yes
Approximate Constant 0.002
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Includes UKAS ISO 17025 calibration
Size 25
8 week lead-time