missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cannabinoid Receptor 1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142759
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize reported isoform a (NP_057167.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.
Specifications
| Cannabinoid Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| CNR1 | |
| brain-type cannabinoid receptor; CANN6; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB1; CB1 cannabinoid receptor; CB1 receptor; CB1A; CB1K5; CB1R; CB-R; central cannabinoid receptor; CNR; Cnr1; HGNC:2159; OTTHUMP00000016839; RP1-23D17.1; seven transmembrane domains; G-protein-coupled receptor; Skr6; SKR6R; striatal cannabinoid receptor type 1 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1268, 12801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P21554, P47746 | |
| CNR1 | |
| Peptide with sequence SNDIQYEDIKGDMAS-C (aa 25-39). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction