missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody, Novus Biologicals™
Description
Cannabinoid R1/CB1/CNR1 Polyclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Cannabinoid R1/CB1/CNR1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR |
| Host Species | Rabbit |
| Immunogen | Synthetic 20 amino acid peptide from N-Terminus extracellular domain of human Cannabinoid R1/CB1/CNR1. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?