missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody (368302), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3834PECY7
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry
Specifications
| Cannabinoid R1/CB1/CNR1 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 368302 | |
| Flow Cytometry | |
| CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
| HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167 | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
| 1268 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction