missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (-)-Cannabidiol

Product Code. 18094992
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18094992

Brand: Tocris Bioscience™ 1570/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Natural cannabinoid; GPR55 antagonist, weak CB1 antagonist, CB2 inverse agonist and AMT inhibitor

(-)-Cannabidiol is a non-psychotropic constituent of cannabis that is anticonvulsive, antihyperalgesic and neuroprotective in vivo. GPR55 and weak CB1 antagonist (IC50 values are 0.445 and 3.35 μM), CB2 receptor inverse agonist and inhibitor of anandamide uptake (IC50 = 27.5 μM). Also inhibits FABPs and a weak agonist at TRP vanilloid receptors (EC50 = 3.5 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol
CAS 13956-29-1
Quantity 10 mg
Target Non-selective Cannabinoid Receptor Antagonists
Molecular Formula C21H30O2
Purity 0.99
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.