missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabidiol, 1mg/mL in Methanol, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Cannabidiol, 1mg/mL in Methanol
Specifications
Specifications
| Chemical Name or Material | Cannabidiol |
| CAS | 13956-29-1 |
| Analyte or Component Names | Cannabidiol |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C21 H30 O2 |
| Concentration | 1 mg/mL |
| Synonym | (-)-Cannabidiol, 2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol, (1R-trans)-2-[3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol, Cannabidiol, trans-(-)-2-p-Mentha-1,8-dien-3-yl-5-pentylresorcinol, (-)-CBD, (-)-trans-Cannabidiol, CBD, Δ1(2)-trans-Cannabidiol, Cannabinoids |
| InChI Formula | InChI=1S/C21H30O2/c1-5-6-7-8-16-12-19(22)21(20(23)13-16)18-11-15(4)9-10-17(18)14(2)3/h11-13,17-18,22-23H,2,5-10H2,1,3-4H3/t17-,18+/m0/s1 |
| SMILES | CCCCCc1cc(O)c([C@@H]2C=C(C)CC[C@H]2C(=C)C)c(O)c1 |
| IUPAC Name | 2-[(1R,6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction