missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine VEGF164 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1603-SP
This item is not returnable.
View return policy
Description
VEGF Polyclonal specifically detects VEGF in Canine samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| VEGF | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7422 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.04-0.10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9MYV3 | |
| VEGFA | |
| E. coli-derived recombinant canine VEGF 164 Pro28-Arg190 Accession # Q9MYV3 | |
| 25 μg | |
| Primary | |
| Detects canine VEGF 164 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human VEGF, recombinant mouse VEGF, and recombinant rat VEGF is observed. | |
| Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction