missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Canine Parvovirus Monoclonal Antibody (A92C)

Description
The MA1-7467 antibody reacts with Canine Parvovirus in viral and infected tissue samples. MA1-7467 has been successfully used in ELISA applications. The MA1-7467 antibody was raised against a parovirus viral preparation.
Canine parvovirus is a contagious virus mainly affecting dogs. CPV is highly contagious and is spread from dog to dog by direct or indirect contact with their feces. Vaccines can prevent this infection, but mortality can reach 91% in untreated cases. Treatment often involves veterinary hospitalization.
Specifications
Specifications
| Antigen | Canine Parvovirus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A92C |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | CPV; parvo |
| Host Species | Mouse |
| Immunogen | Viral preparation. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?