missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-8/CXCL8 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1608
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Polyclonal specifically detects CXCL8/IL-8 in Canine samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| CXCL8/IL-8 | |
| Polyclonal | |
| Biotin | |
| P41324 | |
| IL8 | |
| E. coli-derived recombinant canine IL-8/CXCL8 Ala23-Pro101 Accession # P41324 | |
| 50 μg | |
| Primary | |
| Detects canine IL-8/CXCL8 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human IL-8/CXCL8 and recombinant porcine IL-8/CXCL8 is observed. | |
| Canine | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3576 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction