missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6658-SP
This item is not returnable.
View return policy
Description
IL-3 Monoclonal specifically detects IL-3 in Canine samples. It is validated for Immunocytochemistry.
Specifications
| IL-3 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 20-30 ug/mL | |
| Q9BDX4 | |
| IL3 | |
| E. coli-derived recombinant canine IL-3 Arg24-Pro143 Accession # Q9BDX4 | |
| 25 μg | |
| Primary | |
| Detects canine IL-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant IL-3 from human, mouse, or rat is observed. | |
| Canine | |
| Purified |
| Immunocytochemistry | |
| 662008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3562 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction