missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine IL-21 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5849
This item is not returnable.
View return policy
Description
IL-21 Polyclonal specifically detects IL-21 in Canine samples. It is validated for Neutralization.
Specifications
| IL-21 | |
| Polyclonal | |
| Unconjugated | |
| NP_001003347 | |
| IL21 | |
| E. coli-derived recombinant canine IL-21 His18-Ser146 Accession # NP_001003347 | |
| 100 μg | |
| Primary | |
| Detects canine IL-21 in direct ELISAs. In direct ELISAs, approximately 35% cross-reactivity with recombinant human IL-21 is observed, and less than 3% cross-reactivity with recombinant mouse IL-21 is observed. | |
| Canine | |
| IgG |
| Neutralization | |
| LYOPH | |
| Neutralization 2-10 ug/mL | |
| CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 59067 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction