Learn More
R&D Systems™ Canine IL-17 ELISpot Development Module, 5 Plate
Product Code. 18432430
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18432430 1 Kit 1 set
1 options
Product Code. 18432430 Supplier R&D Systems™ Supplier No. SEL5848
Includes
Canine IL-17/IL-17A Capture Antibody; Canine IL-17/IL-17A Biotinylated-conjugated Detection Antibody
Offers an economical alternative to buying separate antibodies
Contains the basic components required to develop an ELISpot (quantitative sandwich ELISA) assay
Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
Generic development protocols provide directi
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Synonym CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.