Learn More
R&D Systems™ Canine IFN-gamma ELISpot Development Module, 5 Plate
Product Code. 18412410
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18412410 1 Kit 1 set
1 options
Product Code. 18412410 Supplier R&D Systems™ Supplier No. SEL781
Sufficient for at least five 96-well plates
Product Features:
  • An economical alternative to ELISpot Kits
  • Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
  • Generic development protocols provide direction to start an optimization protocol
  • Customize the assay to your specific needs
Module content:
  • Capture Antibody
  • Biotinylated-conjugated Detection Antibody
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.