missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine Distemper Virus Antibody (DV2-12), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64816C
This item is not returnable.
View return policy
Description
Canine Distemper Virus Monoclonal specifically detects Canine Distemper Virus in Canine, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Canine Distemper Virus | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-64816 recognizes canine distemper virus nucleoprotein. In Western blots the antibody recognizes bands of 76kD, 68kD, 54kD and 32kD under reducing conditions. This antibody is useful for the detection of CDV in immunofluorescence procedures, in which the staining seen has a cytoplasmic pattern in infected cells. It is also effective in paraffin embedded material. This antibody is anti morbillivirus specific and reacts with phocine distemper virus, racoon distemper virus and human measles virus. It does not cross react with CAV2, CCV, CPI or CPV. | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DV2-12 | |
| Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CDV | |
| Made to Viral CANINE DISTEMPER VIRUS | |
| 0.125 mL | |
| Primary | |
| Canine, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction