missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canertinib, MedChemExpress
Description
Canertinib (CI-1033
Specifications
Specifications
| Chemical Name or Material | Canertinib |
| CAS | 267243-28-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H25ClFN5O3 |
| Quantity | 200 mg |
| Synonym | CI-1033 PD-183805 |
| Solubility Information | Ethanol : 12.5 mg/mL (25.72 mM; Need ultrasonic) ∣DMSO : 4.9 mg/mL (10.08 mM; Need warming) |
| SMILES | O=C(NC1=C(C=C2C(C(NC3=CC=C(C(Cl)=C3)F)=NC=N2)=C1)OCCCN4CCOCC4)C=C |
| Molecular Weight (g/mol) | 485.94 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?