missing translation for 'onlineSavingsMsg'
Learn More

Castanospermine, MedChemExpress

Product Code. 16474927
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16474927

Brand: Medchem Express HYN2022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Castanospermine inhibits all forms of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids).

Castanospermine inhibits all forms of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids).

Specifications

Chemical Name or Material Castanospermine
CAS 79831-76-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H15NO4
Solubility Information H2O : 100 mg/mL (528.51 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (528.51 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@]([C@@H](O)CC2)([H])N2C1
Molecular Weight (g/mol) 189.21
Quantity 50 mg
Formula Weight 189.21
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.