Learn More
Invitrogen™ Candida albicans Recombinant Rabbit Monoclonal Antibody (6405)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA533304
Description
Reactive with C. albicans (10 strains), C. parapsillosis, C. glabrata, C. tropicales & C. auris. Negative for C. krusei. Non-reactive with: Aspergillus species (4), Coccidioides, Histoplasma, Blastomyces, Paracoccidioides, Penicillium notatum, Mucor, Saccharomyces cerevisiae, and Pichia pastoris.
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).
Specifications
| Candida albicans | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.5 | |
| Rabbit | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 6405 | |
| Unconjugated | |
| C. albicans | |
| Protein A | |
| RUO | |
| Yeast | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.