missing translation for 'onlineSavingsMsg'
Learn More

Candesartan, MedChemExpress

Product Code. 16458418
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458418 10 mg 10mg
16424307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458418 Supplier Medchem Express Supplier No. HYB0205
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Candesartan is an angiotensin II receptor antagonist with IC50 of 0.26 nM.

Candesartan is an angiotensin II receptor antagonist with IC50 of 0.26 nM.

Specifications

Chemical Name or Material Candesartan
CAS 139481-59-7
Color White
Purity Grade Notes Research
Molecular Formula C24H20N6O3
Quantity 10 mg
Synonym CV 11974
Solubility Information DMSO : ≥ 100 mg/mL (227.04 mM)
SMILES O=C(C1=C2C(N=C(OCC)N2CC3=CC=C(C4=CC=CC=C4C5=NNN=N5)C=C3)=CC=C1)O
Molecular Weight (g/mol) 440.45
Formula Weight 440.45
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.