missing translation for 'onlineSavingsMsg'
Learn More

Candesartan-d4, MedChemExpress

Product Code. 16480818
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480818 5 mg 5mg
16468418 10 mg 10mg
16412089 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480818 Supplier Medchem Express Supplier No. HYB0205S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.

Candesartan D4 (CV-11974 D4) is the deuterium labeled Candesartan, which is an angiotensin II receptor antagonist.

Specifications

Chemical Name or Material Candesartan-d4
CAS 1346604-70-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C24H16D4N6O3
Color White
Synonym CV-11974-d4
SMILES [2H]C1=C(CN2C(C(C(O)=O)=CC=C3)=C3N=C2OCC)C([2H])=C([2H])C(C4=CC=CC=C4C5=NNN=N5)=C1[2H]
Molecular Weight (g/mol) 444.48
Quantity 5 mg
Formula Weight 444.48
Percent Purity 98.85%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.