Learn More
Invitrogen™ CAND2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120458
Description
Positive test controls include: HeLa, 293T, Jurkat, NIH/3T3, Mouse heart, Rat heart. The target is usually found in the following locations: Nucleus. Immunogen sequence: VGQVAGPGHQ RELKAVLLEA LGSPSEDVRA AASYALGRVG AGSLPDFLPF LLEQIEAEPR RQYLLLHSLR EALGAAQPDS LKPYAEDIWA LLFQRCEGAE EGTRGVVAEC IGKLVLVNPS FLLPRLRKQL AAGRPHTRST VITAVKFLIS D.
CAND2 may play a role in the assembly of ubiquitin ligase complexes and modulate the ubiquitination of target proteins. And it may be also a transcription regulator.
Specifications
| CAND2 | |
| Polyclonal | |
| Unconjugated | |
| CAND2 | |
| 2210404G23Rik; cand1; Cand2; cullin associated and neddylation dissociated 2 (putative); cullin-associated and neddylation-dissociated 1; cullin-associated and neddylation-dissociated 2 (putative); cullin-associated and neddylation-dissociated protein 2; Cullin-associated NEDD8-dissociated protein 2; epididymis tissue protein Li 169; KIAA0667; mKIAA0667; p120 CAND2; si:ch211-257f16.2; si:dkey-72g22.1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120B; TBP-interacting protein b; TBP-interacting protein of 120 kDa B; TBP-interacting protein Tip120B; Tip120B; Tp120b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192226, 23066, 67088 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.05 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75155, Q6ZQ73, Q9R0L4 | |
| CAND2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.