missing translation for 'onlineSavingsMsg'
Learn More

Canagliflozin hemihydrate, MedChemExpress

Product Code. 16494707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494707 50 mg 50mg
16461238 5 mg 5mg
16428808 10 mg 10mg
16408936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494707 Supplier Medchem Express Supplier No. HYI0383

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Canagliflozin hemihydrate (JNJ28431754 hemihydrate) is a selective SGLT2 inhibitor with IC50s of 2 nM, 3.7 nM, and 4.4 nM for mSGLT2, rSGLT2, and hSGLT2 in CHOK cells, respectively.

Canagliflozin hemihydrate (JNJ28431754 hemihydrate) is a selective SGLT2 inhibitor with IC50s of 2 nM, 3.7 nM, and 4.4 nM for mSGLT2, rSGLT2, and hSGLT2 in CHOK cells, respectively.

Specifications

Chemical Name or Material Canagliflozin hemihydrate
CAS 928672-86-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26FO5.5S
Quantity 50 mg
Synonym JNJ 28431754 hemihydrate
Solubility Information DMSO : ≥ 100 mg/mL (220.50 mM)
SMILES CC1=CC=C([C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)C=C1CC3=CC=C(C4=CC=C(F)C=C4)S3.[0.5H2O]
Molecular Weight (g/mol) 453.52
Formula Weight 453.52
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.