missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canada Pesticide Mixture 3 20-1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Azadirachtin A∣Chlorfenapyr∣Clothianidin∣Cyantraniliprole∣Daminozide∣Dodemorph∣Etridiazole∣Fludioxonil∣Fluopyram∣MGK 264∣Naled∣Parathion-methyl∣Pyrethrins |
| CAS | 11141-17-6, 122453-73-0, 210880-92-5, 736994-63-1, 1596-84-5, 1593-77-7 |
| Analyte or Component Names | Azadirachtin A∣Chlorfenapyr∣Clothianidin∣Cyantraniliprole∣Daminozide∣Dodemorph∣Etridiazole∣Fludioxonil∣Fluopyram∣MGK 264∣Naled∣Parathion-methyl∣Pyrethrins |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Cyantraniliprole 20 μg/ml∣Fludioxonil 20 μg/ml∣Fluopyram 20 μg/ml∣Etridiazole 30 μg/ml∣Chlorfenapyr 50 μg/ml∣Clothianidin 50 μg/ml∣Dodemorph 50 μg/ml∣MGK 264 50 μg/ml∣Parathion-methyl 50 μg/ml∣Pyrethrins 50 μg/ml∣Daminozide 100 μg/ml∣Naled 100 μg/ml∣Azadirachtin A 1000 μg/ml |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?