missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canada Pesticide Mixture 2B 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dimethomorph∣Fenthion∣Imazalil∣Naled∣Parathion-methyl∣Spirodiclofen∣Spiroxamine |
| CAS | 110488-70-5, 55-38-9, 35554-44-0, 300-76-5, 298-00-0, 148477-71-8 |
| Analyte or Component Names | Dimethomorph∣Fenthion∣Imazalil∣Naled∣Parathion-methyl∣Spirodiclofen∣Spiroxamine |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dimethomorph 100 μg/mL∣Fenthion 100 μg/mL∣Imazalil 100 μg/mL∣Naled 100 μg/mL∣Parathion-methyl 100 μg/mL∣Spirodiclofen 100 μg/mL∣Spiroxamine 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction