missing translation for 'onlineSavingsMsg'
Learn More

Canada Pesticide Mixture 2A 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30322890
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322890 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322890 Supplier Dr. Ehrenstorfer Supplier No. DREA50000478AL
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acephate∣Allethrin∣alpha-Endosulfan∣beta-Endosulfan∣Bifenthrin∣Buprofezin∣Chlorfenapyr∣Chlorpyrifos∣Coumaphos∣Cyfluthrin∣Cypermethrin∣Cyprodinil∣Deltamethrin∣Endosulfan-sulfate∣Etofenprox∣Etoxazole∣Etridiazole∣Fenoxycarb∣Fensulfothion∣Fenvalerate∣Fipronil∣Fludioxonil∣Iprodione∣Kinoprene∣Kresoxim-methyl∣Methoprene∣Mevinphos∣MGK 264∣Permethrin∣Phenothrin∣Pirimicarb∣Prallethrin∣Propiconazole∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Tetramethrin∣Trifloxystrobin
CAS 30560-19-1, 584-79-2, 959-98-8, 33213-65-9, 82657-04-3, 69327-76-0
Analyte or Component Names Acephate∣Allethrin∣alpha-Endosulfan∣beta-Endosulfan∣Bifenthrin∣Buprofezin∣Chlorfenapyr∣Chlorpyrifos∣Coumaphos∣Cyfluthrin∣Cypermethrin∣Cyprodinil∣Deltamethrin∣Endosulfan-sulfate∣Etofenprox∣Etoxazole∣Etridiazole∣Fenoxycarb∣Fensulfothion∣Fenvalerate∣Fipronil∣Fludioxonil∣Iprodione∣Kinoprene∣Kresoxim-methyl∣Methoprene∣Mevinphos∣MGK 264∣Permethrin∣Phenothrin∣Pirimicarb∣Prallethrin∣Propiconazole∣Pyraclostrobin∣Pyridaben∣Resmethrin∣Tetramethrin∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acephate 100 μg/ml∣Allethrin 100 μg/ml∣alpha-Endosulfan 100 μg/ml∣beta-Endosulfan 100 μg/ml∣Bifenthrin 100 μg/ml∣Buprofezin 100 μg/ml∣Chlorfenapyr 100 μg/ml∣Chlorpyrifos 100 μg/ml∣Coumaphos 100 μg/ml∣Cyfluthrin 100 μg/ml∣Cypermethrin 100 μg/ml∣Cyprodinil 100 μg/ml∣Deltamethrin 100 μg/ml∣Endosulfan-sulfate 100 μg/ml∣Etofenprox 100 μg/ml∣Etoxazole 100 μg/ml∣Etridiazole 100 μg/ml∣Fenoxycarb 100 μg/ml∣Fensulfothion 100 μg/ml∣Fenvalerate 100 μg/ml∣Fipronil 100 μg/ml∣Fludioxonil 100 μg/ml∣Iprodione 100 μg/ml∣Kinoprene 100 μg/ml∣Kresoxim-methyl 100 μg/ml∣Methoprene 100 μg/ml∣Mevinphos 100 μg/ml∣MGK 264 100 μg/ml∣Permethrin 100 μg/ml∣Phenothrin 100 μg/ml∣Pirimicarb 100 μg/ml∣Prallethrin 100 μg/ml∣Propiconazole 100 μg/ml∣Pyraclostrobin 100 μg/ml∣Pyridaben 100 μg/ml∣Resmethrin 100 μg/ml∣Tetramethrin 100 μg/ml∣Trifloxystrobin 100 μg/ml
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.