missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMTA1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total CAMTA1 protein.
CAMTA1 is a transcriptional activator. May act as a tumor suppressor.
Specifications
Specifications
| Antigen | CAMTA1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CAMTA1 |
| Gene Accession No. | A2A891, Q9Y6Y1 |
| Gene Alias | 1810059M14Rik; 2310058O09Rik; AI316882; AW987475; calmodulin binding transcription activator 1; calmodulin-binding transcription activator 1; Camta1; CANPMR; Kiaa0833; mKIAA0833; MSTP023 |
| Gene Symbols | CAMTA1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?