missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMSAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555301
This item is not returnable.
View return policy
Description
Immunogen sequence: SSVHGVSFDI SFDKEDSVQR STPNRGITRS ISNEGLTLNN SHVSKHIRKN LSFKPINGEE EAESIEEELN IDSHSDLKSC VPL Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.
Specifications
| CAMSAP2 | |
| Polyclonal | |
| Unconjugated | |
| CAMSAP2 | |
| 1600013L13Rik; 4930541M15Rik; calmodulin regulated spectrin associated protein family member 2; calmodulin regulated spectrin-associated protein 1-like 1; calmodulin regulated spectrin-associated protein family, member 2; calmodulin-regulated spectrin-associated protein 1-like protein 1; Calmodulin-regulated spectrin-associated protein 2; CAMSAP1L1; CAMSAP2; KIAA1078; mKIAA1078; RGD1310950 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q08AD1 | |
| CAMSAP2 | |
| Recombinant protein corresponding to Human CAMSAP2. Recombinant protein control fragment (Product #RP-94138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction