missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAMSAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599978
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CAMSAP2.
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.
Specifications
| CAMSAP2 | |
| Polyclonal | |
| Unconjugated | |
| CAMSAP2 | |
| 1600013L13Rik; 4930541M15Rik; calmodulin regulated spectrin associated protein family member 2; calmodulin regulated spectrin-associated protein 1-like 1; calmodulin regulated spectrin-associated protein family, member 2; calmodulin-regulated spectrin-associated protein 1-like protein 1; Calmodulin-regulated spectrin-associated protein 2; CAMSAP1L1; CAMSAP2; KIAA1078; mKIAA1078; RGD1310950 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23271, 289400, 67886 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q08AD1, Q8C1B1 | |
| CAMSAP2 | |
| A synthesized peptide derived from human CAMSAP2(Accession Q08AD1), corresponding to amino acid residues S1290-S1340. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction