missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMSAP2 Mouse anti-Human, Mouse, Rat, Clone: 2A10A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66463-1-IG-150UL
This item is not returnable.
View return policy
Description
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.Specifications
| CAMSAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08AD1, Q8C1B1 | |
| Camsap2 | |
| CAMSAP2 Fusion Protein Ag11794 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2A10A1 | |
| Unconjugated | |
| CAMSAP2 | |
| CAMSAP1L1, CAMSAP2, KIAA1078 | |
| Mouse | |
| Protein G | |
| RUO | |
| 23271, 289400, 67886 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction