missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMSAP1L1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21401JF549
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CAMSAP1L1 Polyclonal antibody specifically detects CAMSAP1L1 in Human samples. It is validated for Western Blot
Specifications
| CAMSAP1L1 | |
| Polyclonal | |
| Western Blot | |
| calmodulin regulated spectrin-associated protein 1-like 1, calmodulin-regulated spectrin-associated protein 2, CAMSAP2, KIAA1078Calmodulin-regulated spectrin-associated protein 1-like protein 1, MGC150680, MGC150681 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human calmodulin regulated spectrin-associated protein 1-like 1 using the numbering given in entry EAW91320.1 (GeneID 23271). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23271 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction