missing translation for 'onlineSavingsMsg'
Learn More

CAMSAP1L1 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18408943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408943 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408943 Supplier Novus Biologicals Supplier No. NBP121401C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CAMSAP1L1 Polyclonal antibody specifically detects CAMSAP1L1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMSAP1L1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias calmodulin regulated spectrin-associated protein 1-like 1, calmodulin-regulated spectrin-associated protein 2, CAMSAP2, KIAA1078Calmodulin-regulated spectrin-associated protein 1-like protein 1, MGC150680, MGC150681
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human calmodulin regulated spectrin-associated protein 1-like 1 using the numbering given in entry EAW91320.1 (GeneID 23271).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23271
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.