missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Campylobacter jejuni Monoclonal Antibody (57-24)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA135036
This item is not returnable.
View return policy
Description
The antibody reacts reacts with a cell wall component of C. jejuni and C. coli. The Hippurate reaction was considered as a marker for C. jejuni. The antibody does not react with C. fetus or C. laridus.
Campylobacter jejuni is a species of curved, long rod-shaped, Gram-negative microaerophilic, bacteria commonly found in animal feces. It is one of the most common causes of human gastroenteritis in the world. C. jejuni is commonly associated with poultry and naturally colonizes the G1 tract of many bird species.
Specifications
| Campylobacter jejuni | |
| Monoclonal | |
| 100 μg/mL | |
| Tissue culture supernatant, PBS with 0.1% BSA and 0.02% sodium azide | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA | |
| 57-24 | |
| Unconjugated | |
| C. jejuni | |
| Proprietary Immunogen. | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction