missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Campylobacter Jejuni Antibody (380/412), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62426AF350
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Campylobacter Jejuni Monoclonal specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA.
Spécification
| Campylobacter Jejuni | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-62426 is specific for Campylobacter jejuni. Some reactivity has been observed with Penner 4 +/- and with Penner 10. C. jejuni is a curved, rod-shaped gram negative bacterium which is a member of the Campylobacteraceae family. They are motile with a polar unsheathed flagellum that results in a corkscrew like motion. They occur primarily as commensals in humans and domestic animals, particularly in chickens where gut temperatures of 42oC are optimal for growth. C. jejuni are one of the most common causes of human diarrhea in the world. Food poisoning can be severely debilitating, although rarely life-threatening as the disease is self limiting. Campylobacter infections are also associated with postinfectious complications including arthritis, Reiter syndrome and GuillainûBarre┤ syndrome. | |
| Store at 4°C in the dark. |
| ELISA | |
| 380/412 | |
| ELISA | |
| Campilobacter | |
| C. jejuni Penner 2 | |
| Primary | |
| Bacteria | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu